Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-3-cyclohexyl-L-alanine 98.0+%, TCI America™
SDP

Catalog No. F08571G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F08571G 1 g
F08575G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F08571G Supplier TCI America Supplier No. F08571G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 135673-97-1
Molecular Formula C24H27NO4
Molecular Weight (g/mol) 393.48
MDL Number MFCD00065614
InChI Key HIJAUEZBPWTKIV-UHFFFAOYNA-N
Synonym fmoc-cha-oh, fmoc-3-cyclohexyl-l-alanine, fmoc-l-cyclohexylalanine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-cyclohexylpropanoic acid, n-fmoc-3-cyclohexyl-l-alanine, n-9h-fluoren-9-ylmethoxy carbonyl-3-cyclohexyl-l-alanine, n-alpha-9-fluorenylmethyloxycarbonyl-l-cyclohexylalanine, 2s-3-cyclohexyl-2-9h-fluoren-9-ylmethoxycarbonylamino propanoic acid, 2s-3-cyclohexyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, fmoc-3-cyclohexylalanine
PubChem CID 978326
IUPAC Name 3-cyclohexyl-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid
SMILES OC(=O)C(CC1CCCCC1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Melting Point 140°C
Color White
Quantity 1 g
Formula Weight 393.48
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-3-cyclohexyl-L-alanine
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.