Learn More
Medchemexpress LLC N-[(9H-fluoren-9-ylmethoxy)carbonyl]-3-methoxy-L-phenylalanine | 206060-40-4 | MFCD01317009 | 99.6% | 417.45 g/mol | C25H23NO5 | 10 G
SDP
Catalog No. 5000300118
Encompass_Preferred
N-[(9H-fluoren-9-ylmethoxy)carbonyl]-3-methoxy-L-phenylalanine is an Fmoc-protected derivative of L-phenylalanine bearing a methoxy group at the 3-position of the aromatic ring. Supplied as a high-purity solid, it serves as a protected amino acid building block for Fmoc-based solid-phase peptide synthesis and for incorporation of a 3-methoxyphenylalanine residue into modified peptides.
  • High reported purity suitable for peptide synthesis.
  • Molecular weight approximately 417.45 g/mol.
  • Solid form for convenient handling and storage.
  • Compatible with standard Fmoc-based SPPS protocols.
  • Enables incorporation of a 3-methoxy substitution into peptide sequences.
Catalog No. 50-003-00118 Supplier Medchemexpress LLC Supplier No. HYW02222810G
N-[(9H-fluoren-9-ylmethoxy)carbonyl]-3-methoxy-L-phenylalanine is an Fmoc-protected derivative of L-phenylalanine bearing a methoxy group at the 3-position of the aromatic ring. Supplied as a high-purity solid, it serves as a protected amino acid building block for Fmoc-based solid-phase peptide synthesis and for incorporation of a 3-methoxyphenylalanine residue into modified peptides.
  • High reported purity suitable for peptide synthesis.
  • Molecular weight approximately 417.45 g/mol.
  • Solid form for convenient handling and storage.
  • Compatible with standard Fmoc-based SPPS protocols.
  • Enables incorporation of a 3-methoxy substitution into peptide sequences.