Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-aspartic Acid 98.0+%, TCI America™
SDP

Catalog No. p-7129773 Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F05921G 1 g
F05925G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F05921G Supplier TCI America Supplier No. F05921G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 136083-57-3
Molecular Formula C19H17NO6
Molecular Weight (g/mol) 355.346
MDL Number MFCD01318740
InChI Key KSDTXRUIZMTBNV-MRXNPFEDSA-N
Synonym N-Fmoc-D-aspartic Acid, Fmoc-D-Asp-OH
PubChem CID 7006674
IUPAC Name (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)butanedioic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC(=O)O)C(=O)O

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 355.35
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-aspartic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.