Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-glutamic Acid 97.0+%, TCI America™
SDP

Catalog No. F06005G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F06005G 5 g
F060025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F06005G Supplier TCI America Supplier No. F06005G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 104091-09-0
Molecular Formula C20H19NO6
Molecular Weight (g/mol) 369.373
InChI Key QEPWHIXHJNNGLU-QGZVFWFLSA-N
Synonym N-Fmoc-D-glutamic Acid, Fmoc-D-Glu-OH
PubChem CID 40501223
IUPAC Name (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)pentanedioic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CCC(=O)O)C(=O)O

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 369.37
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-glutamic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.