Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-threonine 98.0+%, TCI America™
SDP

Catalog No. F06085G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F06085G 5 g
F06081G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F06085G Supplier TCI America Supplier No. F06085G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 157355-81-2
Molecular Formula C19H19NO5
Molecular Weight (g/mol) 341.363
InChI Key OYULCCKKLJPNPU-APPDUMDISA-N
Synonym N-Fmoc-D-threonine, Fmoc-D-Thr-OH
PubChem CID 6992532
IUPAC Name (2R,3S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-hydroxybutanoic acid
SMILES CC(C(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13)O

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 341.36
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-threonine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.