Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-2-phenylglycine 98.0+%, TCI America™
SDP

Catalog No. F06695G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F06695G 5 g
F06691G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F06695G Supplier TCI America Supplier No. F06695G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 102410-65-1
Molecular Formula C23H19NO4
Molecular Weight (g/mol) 373.408
MDL Number MFCD00155632
InChI Key PCJHOCNJLMFYCV-NRFANRHFSA-N
Synonym fmoc-phg-oh, fmoc-l-phenylglycine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-2-phenylacetic acid, n-fmoc-l-2-phenylglycine, fmoc-s-phenylglycine, n-9h-fluoren-9-ylmethoxy carbonyl-l-2-phenylglycine, n-alpha-9-fluorenylmethyloxycarbonyl-l-phenylglycine, s-9h-fluoren-9-ylmethoxycarbonylamino-phenyl-acetic acid, s-9h-fluoren-9-ylmethoxy carbonyl amino phenyl acetic acid, 2s-9h-fluoren-9-yl-methoxy carbonyl amino phenyl acetic acid
PubChem CID 7269367
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-2-phenylacetic acid
SMILES C1=CC=C(C=C1)C(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24

Specifications

Melting Point 176°C
Color White-Yellow
Quantity 5 g
Formula Weight 373.41
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-2-phenylglycine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.