Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-isoleucine, TCI America™
SDP

Catalog No. F02945G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F02945G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F02945G Supplier TCI America Supplier No. F02945G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 71989-23-6
Molecular Formula C21H23NO4
Molecular Weight (g/mol) 353.42
MDL Number MFCD00037125
InChI Key QXVFEIPAZSXRGM-DJJJIMSYSA-N
Synonym fmoc-ile-oh, fmoc-l-isoleucine, n-fmoc-l-isoleucine, fmoc-l-ile-oh, n-9h-fluoren-9-ylmethoxy carbonyl-l-isoleucine, n-9-fluorenylmethoxycarbonyl-l-isoleucine, fmoc-ile, fmoc-l-ile, 2s,3s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-methylpentanoic acid, 9-fluorenylmethoxycarbonyl-l-isoleucine
PubChem CID 2724629
IUPAC Name (2S,3S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-methylpentanoic acid
SMILES CC[C@H](C)[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

Melting Point 146°C
Color White
Quantity 5 g
Formula Weight 353.42
Physical Form Crystalline Powder
Chemical Name or Material N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-isoleucine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.