Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-phenylalanine 98.0+%, TCI America™
SDP

Catalog No. F02975G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F02975G 5 g
F029725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F02975G Supplier TCI America Supplier No. F02975G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 35661-40-6
Molecular Formula C24H21NO4
Molecular Weight (g/mol) 387.44
MDL Number MFCD00037128
InChI Key SJVFAHZPLIXNDH-QFIPXVFZSA-N
Synonym fmoc-l-phenylalanine, fmoc-phe-oh, n-fmoc-l-phenylalanine, n-9-fmoc-l-phenylglycine, n-9-fluorenylmethoxycarbonyl-l-phenylalanine, fmoc-l-phe-oh, fmoc-phe, fmoc-l-phe, fmoc-phenylalanine, n-9h-fluoren-9-ylmethoxy carbonyl-l-phenylalanine
PubChem CID 978331
IUPAC Name (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-phenylpropanoic acid
SMILES OC(=O)[C@H](CC1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Melting Point 183°C
Color White
Quantity 5 g
Formula Weight 387.44
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-phenylalanine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.