Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-serine Hydrate 97.0+%, TCI America™
SDP

Catalog No. F04545G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F04545G 5 g
F04541G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F04545G Supplier TCI America Supplier No. F04545G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 73724-45-5
Molecular Formula C18H16NO5
Molecular Weight (g/mol) 326.33
MDL Number MFCD00051928
InChI Key JZTKZVJMSCONAK-INIZCTEOSA-M
Synonym fmoc-ser-oh, fmoc-l-serine, fmoc-l-ser-oh, n-fmoc-l-serine, n-9-fluorenylmethoxycarbonyl-l-serine, n-9h-fluoren-9-ylmethoxy carbonyl-l-serine, n-9h-fluoren-9-ylmethoxycarbonyl serine, s-fmoc-2-amino-3-hydroxypropionic acid, n-alpha-9-fluorenylmethyloxycarbonyl-l-serine, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-hydroxypropanoic acid
PubChem CID 6541433
IUPAC Name (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-hydroxypropanoate
SMILES OC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C([O-])=O

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 327.34
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-serine Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.