Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-methyl-L-norvaline 98.0+%, TCI America™
SDP

Catalog No. F11661G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F11661G 1 g
F11665G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F11661G Supplier TCI America Supplier No. F11661G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 252049-05-1
Molecular Formula C21H23NO4
Molecular Weight (g/mol) 353.42
MDL Number MFCD01861326
InChI Key HKELUUGCKFRJQM-IBGZPJMESA-N
Synonym fmoc-n-methyl-l-norvaline, fmoc-n-me-nva-oh, s-2-9h-fluoren-9-yl methoxy carbonyl methyl amino pentanoic acid, fmoc-n-me-l-2-aminovaleric acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl methyl amino pentanoic acid, l-norvaline, n-9h-fluoren-9-ylmethoxy carbonyl-n-methyl, l-norvaline,n-9h-fluoren-9-ylmethoxy carbonyl-n-methyl, fmoc-n-me-l-norvaline, n-fmoc-n-methyl-l-norvaline, fmoc-nalpha-methyl-l-norvaline
PubChem CID 7009909
IUPAC Name (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}(methyl)amino)pentanoic acid
SMILES CCC[C@H](N(C)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

Melting Point 132°C
Color White
Quantity 1 g
Formula Weight 353.42
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-methyl-L-norvaline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.