Learn More
Medchemexpress LLC N-[(9H-fluoren-9-ylmethoxy)carbonyl]-O-(phenylmethyl)-D-serine | 122889-11-6 | MFCD00237032 | 98.1% | 417.45 g·mol⁻¹ | C25H23NO5 | 25 G
SDP
Catalog No. 5000303037
Encompass_Preferred
N-[(9H-fluoren-9-ylmethoxy)carbonyl]-O-(phenylmethyl)-D-serine is an Fmoc-protected, O-benzylated derivative of D-serine used as a protected amino acid building block for solid-phase peptide synthesis and related organic synthesis applications.
  • Fmoc-protected D-serine with O-benzyl side-chain protection.
  • Suitable for solid-phase peptide synthesis and amino acid coupling.
  • High purity for research and development use.
  • White to off-white solid with stable handling properties.
  • Available in multiple pack sizes for lab-scale use.
Catalog No. 50-003-03037 Supplier Medchemexpress LLC Supplier No. 25GHYW022226
N-[(9H-fluoren-9-ylmethoxy)carbonyl]-O-(phenylmethyl)-D-serine is an Fmoc-protected, O-benzylated derivative of D-serine used as a protected amino acid building block for solid-phase peptide synthesis and related organic synthesis applications.
  • Fmoc-protected D-serine with O-benzyl side-chain protection.
  • Suitable for solid-phase peptide synthesis and amino acid coupling.
  • High purity for research and development use.
  • White to off-white solid with stable handling properties.
  • Available in multiple pack sizes for lab-scale use.