Learn More
Medchemexpress LLC N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-O-(phenylmethyl)-D-serine | 122889-11-6 | MFCD00237032 | 99.9% | 417.45 | C25H23NO5 | 100 G
SDP
Catalog No. 5000301579
Encompass_Preferred
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-O-(phenylmethyl)-D-serine is an Fmoc-protected D-serine derivative with O-benzyl side-chain protection supplied as a solid reagent for research use. It serves as a protected amino acid building block in peptide synthesis and organic synthesis workflows requiring temporary N-protection and side-chain protection.
  • Protected amino acid for peptide synthesis.
  • Fmoc N-protection with O-benzyl side-chain protection.
  • High purity suitable for research use.
  • Solid form with defined storage recommendations.
  • Compatible with standard coupling and deprotection chemistries.
Catalog No. 50-003-01579 Supplier Medchemexpress LLC Supplier No. HYW022226100G
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-O-(phenylmethyl)-D-serine is an Fmoc-protected D-serine derivative with O-benzyl side-chain protection supplied as a solid reagent for research use. It serves as a protected amino acid building block in peptide synthesis and organic synthesis workflows requiring temporary N-protection and side-chain protection.
  • Protected amino acid for peptide synthesis.
  • Fmoc N-protection with O-benzyl side-chain protection.
  • High purity suitable for research use.
  • Solid form with defined storage recommendations.
  • Compatible with standard coupling and deprotection chemistries.