Learn More
Cayman Chemical N-α-Benzoyl-L-Arginine ethyl ester (hydrochloride), Purity: ≥98%, CAS: 2645-08-1, 5 g
SDP
Catalog No. 502938323
Encompass_Preferred
A nonpeptide substrate of proteases and deiminases; has been used as a substrate for the relative quantification of trypsin activity; has also been used as a substrate for the activity of subtilisins, kallikreins, and PAD4
Catalog No. 50-293-8323 Supplier Cayman Chemical Supplier No. 271835 G
A nonpeptide substrate of proteases and deiminases; has been used as a substrate for the relative quantification of trypsin activity; has also been used as a substrate for the activity of subtilisins, kallikreins, and PAD4