Learn More
Cayman Chemical N-a-Benzoyl-L-ArgInInamIdeh 1g
SDP
Supplier:  Cayman Chemical 350151 G
Encompass_Preferred
A nonpeptide substrate of proteases and deiminases; has been used as a substrate for the quantification of papain, carboxypeptidase B, PAD2, and PAD4 activities
Catalog No. 50-291-2307