Learn More
Cayman Chemical N-a-Benzoyl-L-ArgInInamIdeh 5g
SDP
Catalog No. 502911351
Encompass_Preferred
A nonpeptide substrate of proteases and deiminases; has been used as a substrate for the quantification of papain, carboxypeptidase B, PAD2, and PAD4 activities
Catalog No. 50-291-1351 Supplier Cayman Chemical Supplier No. 350155 G
A nonpeptide substrate of proteases and deiminases; has been used as a substrate for the quantification of papain, carboxypeptidase B, PAD2, and PAD4 activities