Learn More
Medchemexpress LLC N-ac-γ-calicheamicin-acbut-NHS ester | 174885-02-0 | 95.6% | 1779.73 | C77H99IN6O28S3 | 1 MG
SDP
Catalog No. 5000246216
Encompass_Preferred
N-Ac-γ-Calicheamicin-AcBut-NHS ester is an activated enediyne-derived cytotoxic payload used for synthesis of antibody-drug conjugates (ADCs) and related research applications. It is supplied as a white to off-white solid with manufacturer-reported analytical data supporting its use in conjugation chemistry.
  • Activated NHS ester for efficient conjugation to primary amines
  • Potent DNA-cleaving enediyne payload suitable for ADC development
  • Supplied as a solid with documented purity and analytical characterization
  • Storage recommendation: -20°C; in solvent stability -80°C for up to 6 months
  • Analytical documentation available (COA, HNMR, RP-HPLC, LC-MS, SDS)
Catalog No. 50-002-46216 Supplier Medchemexpress LLC Supplier No. HY1036881MG
Add to Cart
Add to Cart
Request a Quote
N-Ac-γ-Calicheamicin-AcBut-NHS ester is an activated enediyne-derived cytotoxic payload used for synthesis of antibody-drug conjugates (ADCs) and related research applications. It is supplied as a white to off-white solid with manufacturer-reported analytical data supporting its use in conjugation chemistry.
  • Activated NHS ester for efficient conjugation to primary amines
  • Potent DNA-cleaving enediyne payload suitable for ADC development
  • Supplied as a solid with documented purity and analytical characterization
  • Storage recommendation: -20°C; in solvent stability -80°C for up to 6 months
  • Analytical documentation available (COA, HNMR, RP-HPLC, LC-MS, SDS)