Learn More
Cayman Chemical N-acetyl-2-AmInophenol 50g
SDP
Catalog No. 502911318
Encompass_Preferred
A derivative of acetaminophen; inhibits ADP- or arachidonic acid-induced platelet aggregation in isolated human platelet-rich plasma at 50 µM; decreases disease severity and prevents increases in paw volume in a rat model of collagen-induced arthritis at 1 mg/kg; has been used as a synthetic intermediate in the synthesis of compounds with antimalarial activity; a potential impurity in commercial preparations of acetaminophen
Catalog No. 50-291-1318 Supplier Cayman Chemical Supplier No. 3582950 G
A derivative of acetaminophen; inhibits ADP- or arachidonic acid-induced platelet aggregation in isolated human platelet-rich plasma at 50 µM; decreases disease severity and prevents increases in paw volume in a rat model of collagen-induced arthritis at 1 mg/kg; has been used as a synthetic intermediate in the synthesis of compounds with antimalarial activity; a potential impurity in commercial preparations of acetaminophen