Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-acetyl-2'-deoxy-2'-fluorocytidine | 159414-97-8 | MFCD15145138 | 99.6% | 287.24 g·mol⁻¹ | C11H14FN3O5 | 250 MG
SDP

Catalog No. 5000283268
Encompass_Preferred

N-acetyl-2'-deoxy-2'-fluorocytidine is a purified nucleoside analog used as a building block for oligoribonucleotide synthesis and as a reference compound in nucleoside research.

  • Nucleoside analog suitable for oligoribonucleotide synthesis.
  • High purity (99.6%).
  • Molecular formula C11H14FN3O5 and molecular weight 287.24 g·mol⁻¹.
  • CAS number 159414-97-8.
  • Available in laboratory pack sizes from 250 MG to 5 G.
  • Supplied for biochemical and molecular biology applications as a reference standard.

Catalog No. 50-002-83268 Supplier Medchemexpress LLC Supplier No. HYW131753250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-acetyl-2'-deoxy-2'-fluorocytidine is a purified nucleoside analog used as a building block for oligoribonucleotide synthesis and as a reference compound in nucleoside research.

  • Nucleoside analog suitable for oligoribonucleotide synthesis.
  • High purity (99.6%).
  • Molecular formula C11H14FN3O5 and molecular weight 287.24 g·mol⁻¹.
  • CAS number 159414-97-8.
  • Available in laboratory pack sizes from 250 MG to 5 G.
  • Supplied for biochemical and molecular biology applications as a reference standard.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.