Learn More
Medchemexpress LLC N-acetyl-3-methyl-L-valyl-4-hydroxy-N-(4-(4-methyl-1,3-thiazol-5-yl)benzyl)-L-prolinamide | 1448188-62-2 | 99.6% | 100 MG
SDP
Catalog No. 5000234504
Encompass_Preferred
VH032 is a small-molecule ligand of the von Hippel-Lindau (VHL) E3 ubiquitin ligase used as a VHL-recruiting warhead in proteolysis-targeting chimera (PROTAC) research and to study VHL/HIF-1α interactions.
  • Used as a VHL-recruiting warhead in PROTAC design and targeted protein degradation studies.
  • High chemical purity of 99.6% suitable for biological experiments.
  • Molecular weight 472.6 g·mol⁻¹ and molecular formula C24H32N4O4S.
  • Supplied as a 100 mg solid for synthesis and assay development.
  • Indexed in major chemical databases for reliable compound identification.
Catalog No. 50-002-34504 Supplier Medchemexpress LLC Supplier No. HY120217100MG
VH032 is a small-molecule ligand of the von Hippel-Lindau (VHL) E3 ubiquitin ligase used as a VHL-recruiting warhead in proteolysis-targeting chimera (PROTAC) research and to study VHL/HIF-1α interactions.
  • Used as a VHL-recruiting warhead in PROTAC design and targeted protein degradation studies.
  • High chemical purity of 99.6% suitable for biological experiments.
  • Molecular weight 472.6 g·mol⁻¹ and molecular formula C24H32N4O4S.
  • Supplied as a 100 mg solid for synthesis and assay development.
  • Indexed in major chemical databases for reliable compound identification.