Learn More
Medchemexpress LLC HY-107854 50mg Medchemexpress, N-Acetyl-5-hydroxytryptamine CAS:1210-83-9 Purity:>98%
SDP
Catalog No. 501932302
Encompass_Preferred
Medchemexpress, HY-107854 50mg N-Acetyl-5-hydroxytryptamine CAS:1210-83-9 N-Acetyl-5-hydroxytryptamine is a Melatonin precursor, and that it can potently activate TrkB receptor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2302 Supplier Medchemexpress LLC Supplier No. HY10785450MG
Medchemexpress, HY-107854 50mg N-Acetyl-5-hydroxytryptamine CAS:1210-83-9 N-Acetyl-5-hydroxytryptamine is a Melatonin precursor, and that it can potently activate TrkB receptor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.