Learn More
Selleck Chemical LLC N-Acetyl-5-hydroxytryptamine S2985-5mg
SDP
Catalog No. 503112281
Encompass_Preferred
N-Acetyl-5-hydroxytryptamine (NAS N-Acetylserotonin Normelatonin O-Demethylmelatonin) a precursor of melatonin is a potent and selective agonist of TrkB receptor
Catalog No. 50-311-2281 Supplier Selleck Chemical LLC Supplier No. S29855MG
N-Acetyl-5-hydroxytryptamine (NAS N-Acetylserotonin Normelatonin O-Demethylmelatonin) a precursor of melatonin is a potent and selective agonist of TrkB receptor