Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Acetyl-5-hydroxytryptamine 98.0+%, TCI America™
SDP

Catalog No. A1277100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1277100MG 100 mg
A12771G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A1277100MG Supplier TCI America Supplier No. A1277100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1210-83-9
Molecular Formula C12H14N2O2
Molecular Weight (g/mol) 218.26
MDL Number MFCD00005656
InChI Key MVAWJSIDNICKHF-UHFFFAOYSA-N
Synonym n-acetyl-5-hydroxytryptamine, n-acetylserotonin, n-acetyl serotonin, normelatonin, n-2-5-hydroxy-1h-indol-3-yl ethyl acetamide, o-demethylmelatonin, 5-hydroxy-n-acetyltryptamine, acetylserotonin, unii-p4to3c82wv, 5-hydroxymelatonin
PubChem CID 903
ChEBI CHEBI:17697
IUPAC Name N-[2-(5-hydroxy-1H-indol-3-yl)ethyl]acetamide
SMILES CC(=O)NCCC1=CNC2=CC=C(O)C=C12

Specifications

Melting Point 120°C
Color White-Yellow
Quantity 100 mg
Formula Weight 218.26
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N-Acetyl-5-hydroxytryptamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.