Learn More
CPC Scientific Ac-Ser-Tyr-Ser-Met-Glu-His-Phe-Arg-Trp-Gly-Lys-Pro-Val-Gly-Lys-Lys-Arg-OH (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific ACTH012A
Encompass
ONE-LETTER SEQUENCE: Ac-SYSMEHFRWGKPVGKKRMOLECULAR FORMULA: C97H147N29O24S1MOLECULAR WEIGHT:2135.5STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [197906-63-1], RESEARCH AREA: Hormonal
Catalog No. 50-210-8614