Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-Acetyl-β-alanine | 3025-95-4 | 98.0% | C5H9NO3 | 100 G
SDP

Catalog No. 5000433797
Encompass_Preferred

N-Acetyl-β-alanine (Ac-β-Ala-OH) is an abnormal amino acid metabolite and a mono-N-protected amino acid (MPAA) ligand. It is supplied as a white to off-white solid with a molecular weight of 131.13. This product is intended for research use only and not for patient use.

  • Functions as an abnormal amino acid metabolite
  • Acts as a mono-N-protected amino acid (MPAA) ligand
  • Purity of 98.0%
  • Chemical formula: C5H9NO3
  • Soluble in DMSO at 100 mg/mL

Catalog No. 50-004-33797 Supplier Medchemexpress LLC Supplier No. HY66007100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Acetyl-β-alanine (Ac-β-Ala-OH) is an abnormal amino acid metabolite and a mono-N-protected amino acid (MPAA) ligand. It is supplied as a white to off-white solid with a molecular weight of 131.13. This product is intended for research use only and not for patient use.

  • Functions as an abnormal amino acid metabolite
  • Acts as a mono-N-protected amino acid (MPAA) ligand
  • Purity of 98.0%
  • Chemical formula: C5H9NO3
  • Soluble in DMSO at 100 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.