Learn More
CPC Scientific Ac-Cys-His-Ser-Gly-Tyr-Val-Gly-Val-Arg-Cys-OMe (trifluoroacetate salt) 1MG
SDP
Catalog No. 502109405
Encompass
ONE-LETTER SEQUENCE: Ac-CHSGYVGVRC-Ome (Cys1 and 10 bridge)MOLECULAR FORMULA: C47H72N15O15S2MOLECULAR WEIGHT:1151.3STORAGE CONDITIONS: -20 5CRESEARCH AREA: CancerREFERENCES: Nestor et al., BBRC, 129, 226 (1985)
Catalog No. 50-210-9405 Supplier CPC Scientific Supplier No. GRFF007B
ONE-LETTER SEQUENCE: Ac-CHSGYVGVRC-Ome (Cys1 and 10 bridge)MOLECULAR FORMULA: C47H72N15O15S2MOLECULAR WEIGHT:1151.3STORAGE CONDITIONS: -20 5CRESEARCH AREA: CancerREFERENCES: Nestor et al., BBRC, 129, 226 (1985)