Learn More
Enzo Life Sciences N-Acetyl-Asp-Glu-OH (50mg). CAS: 3106-85-2
Supplier: Enzo Life Sciences ALX151020M050
Glutamate receptor ligand. Endogenous neurotransmitter with high affinity for the brain glutamate receptor. N-acetylated α-linked acidic dipeptidase (NAALADase), a membrane-bound peptidase, hydrolyzes α-NAAG, a major brain peptide, to N-acetylaspartate and glutamate. Discriminates between mGluR2 and mGluR3. mGluR3 activator. NMDA receptor antagonist. Neuroprotective. Alternative name: alpha NAAG, Spaglumic acid. Formula: C11H16N2O8. MW: 304.3. Purity: ≥98% (TLC). Appearance: White to off-white powder. Solubility: Soluble in water and methanol (50mg/ml). Long Term Storage: +4°C.