Learn More
Medchemexpress LLC N-Acetyl-Asp-Glu-Val-Asp-7-amido-4-Methylcoumarin | 169332-61-0 | 99.2% | 675.64 | 25 MG
SDP
Catalog No. 5000425957
Encompass_Preferred
Ac-DEVD-AMC is a fluorescent substrate for caspase-3 and caspase-7. Upon treatment with cell lysate, it facilitates the release of amino-4-methylcoumarin (AMC), which allows for fluorescence detection at an excitation wavelength of 380 nm and an emission wavelength of 460 nm.
  • Fluorescent substrate of caspase-3/caspase-7
  • Releases amino-4-methylcoumarin (AMC) for fluorescence detection
  • Excitation wavelength of 380 nm
  • Emission wavelength of 460 nm
Catalog No. 50-004-25957 Supplier Medchemexpress LLC Supplier No. 25MGHYP1003
Ac-DEVD-AMC is a fluorescent substrate for caspase-3 and caspase-7. Upon treatment with cell lysate, it facilitates the release of amino-4-methylcoumarin (AMC), which allows for fluorescence detection at an excitation wavelength of 380 nm and an emission wavelength of 460 nm.
  • Fluorescent substrate of caspase-3/caspase-7
  • Releases amino-4-methylcoumarin (AMC) for fluorescence detection
  • Excitation wavelength of 380 nm
  • Emission wavelength of 460 nm