Learn More
CPC Scientific Ac-Asp-Tyr(SO3H)-Met-Gly-Trp-NH2 (ammonium salt) 1MG
SDP
Supplier:  CPC Scientific CCKS004B
Encompass
SEQUENCE: Ac-Asp-Tyr(SO3H)-Met-Gly-Trp-NH2 (ammonium salt)ONE-LETTER SEQUENCE: Ac-D-Tyr(SO3H)-MGW-NH2MOLECULAR FORMULA: C33H41N7O12S2MOLECULAR WEIGHT: 791.9STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: SYNONYMS: RESEARCH AREA: GastrointestinalREFERENCES:SKU(s): CCKS-004A, CCKS-004B
Catalog No. 50-210-9116