Learn More
CPC Scientific Ac-Asp-Tyr-Met-Gly-Trp-Met-NH2 (acetate salt) 0.5MG
SDP
Supplier:  CPC Scientific CCKS006A
Encompass
 SEQUENCE: Ac-Asp-Tyr-Met-Gly-Trp-Met-NH2 (acetate salt)ONE-LETTER SEQUENCE: Ac-DYMGWM-NH2MOLECULAR FORMULA: C38H50N8O10S2MOLECULAR WEIGHT: 842.98STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [89911-64-8]SYNONYMS: CCK (26-31) (non-sulfated), cholecystokinin N-acetyl fragment 26-31 amide, non-sulfatedRESEARCH AREA: GastrointestinalREFERENCES:SKU(s): CCKS-006A, CCKS-006B
Catalog No. 50-210-9119