Learn More
Medchemexpress LLC N-acetyl-D-erythro-sphingosine | 3102-57-6 | MFCD00153903 | 99.9% | 341.53 g·mol⁻¹ | C20H39NO3 | 100 MG
SDP
Catalog No. 5000211644
Encompass_Preferred
C2 ceramide (N-acetyl-D-erythro-sphingosine, CAS 3102-57-6) is a short-chain ceramide used in cellular and biochemical research. It functions as a protein phosphatase activator and modulates signaling pathways involved in differentiation, autophagy, and apoptosis. Supplied as a solid suitable for dissolution in organic solvents.
  • Activates protein phosphatases involved in cell regulation.
  • Modulates differentiation, autophagy, and apoptosis.
  • Inhibits mitochondrial respiratory complex III in some studies.
  • Acts as a skin-conditioning agent that supports epidermal barrier function.
  • High purity suitable for research applications.
Catalog No. 50-002-11644 Supplier Medchemexpress LLC Supplier No. HY101180100MG
C2 ceramide (N-acetyl-D-erythro-sphingosine, CAS 3102-57-6) is a short-chain ceramide used in cellular and biochemical research. It functions as a protein phosphatase activator and modulates signaling pathways involved in differentiation, autophagy, and apoptosis. Supplied as a solid suitable for dissolution in organic solvents.
  • Activates protein phosphatases involved in cell regulation.
  • Modulates differentiation, autophagy, and apoptosis.
  • Inhibits mitochondrial respiratory complex III in some studies.
  • Acts as a skin-conditioning agent that supports epidermal barrier function.
  • High purity suitable for research applications.