Learn More
TARGETMOL CHEMICALS INC N-Acetyl-D-galactosamine 25MG
SDP
Catalog No. 502880120
Encompass_Preferred
Also available in 5 mg, 10 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. N-Acetyl-D-galactosamine (GalNAc) is a terminal essential amino sugar derived from galactose and forms the antigens of blood group A in humans. Purity 99.92%
Catalog No. 50-288-0120 Supplier TARGETMOL CHEMICALS INC Supplier No. T588525MG
Also available in 5 mg, 10 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. N-Acetyl-D-galactosamine (GalNAc) is a terminal essential amino sugar derived from galactose and forms the antigens of blood group A in humans. Purity 99.92%