Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Acetyl-D-Galactosamine, MP Biomedicals™

Catalog No. MP21000641 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP21000641 100 mg
MP21000645 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP21000641 Supplier MP Biomedicals, Inc Supplier No. 02100064.1
Add to Cart
Add to Cart

A component of many O-linked and N-linked glycan structures. N-Acetyl-D-Galactosamine, MP Biomedicals is an aminosugar that is the initial O-linked sugar to many serine and threonine residues in protein glycosylations.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14215-68-0
Molecular Formula C8H15NO6
Molecular Weight (g/mol) 221.209
InChI Key OVRNDRQMDRJTHS-JAJWTYFOSA-N
Synonym n-acetyl-d-galactosamine, galnac, 2-acetamido-2-deoxy-d-galactopyranose, d-galnac, 2-acetamido-2-deoxy-d-galactose, n-acetyl-d-chondrosamine, n-3r,4r,5r,6r-2,4,5-trihydroxy-6-hydroxymethyl tetrahydro-2h-pyran-3-yl acetamide, n-acetylgalactosamine, acetylgalactosamine, n-acetyl-galactosamine
PubChem CID 440552
ChEBI CHEBI:28497
IUPAC Name N-[(2R,3R,4R,5R,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide
SMILES CC(=O)NC1C(C(C(OC1O)CO)O)O

Specifications

Melting Point 160°C to 161°C (literature)
Quantity 100 mg
Solubility Information Soluble in water (100mg/mL - clear, colorless solution).
Optical Rotation +84–89° (c=1, water)
Formula Weight 221.21
Physical Form Powder
Recommended Storage Store at +4°C
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.