Learn More
Cayman Chemical N-acetyl-D-Glucosamine, Purity: ≥98%, CAS: 7512-17-6, 500 mg
SDP
Catalog No. 502951719
Encompass_Preferred
A monosaccharide derivative of glucose that is released by the action of O-GlcNAcase, in mammalian systems, from proteins that have been post-translationally modified with O-GlcNAc; also is the monomeric unit of chitin, which is found in fungi and many inverebrates, including crustaceans, insects, and nematodes
Catalog No. 50-295-1719 Supplier Cayman Chemical Supplier No. 13136500 MG
A monosaccharide derivative of glucose that is released by the action of O-GlcNAcase, in mammalian systems, from proteins that have been post-translationally modified with O-GlcNAc; also is the monomeric unit of chitin, which is found in fungi and many inverebrates, including crustaceans, insects, and nematodes