Learn More
Medchemexpress LLC N-acetyl-α-d-glucosamine | 10036-64-3 | MFCD00064359 | 99.4% | 221.21 g/mol | C8H15NO6 | 1 G
SDP
Catalog No. 5000282239
Encompass_Preferred
N-Acetyl-α-D-glucosamine is an amino sugar used as a biochemical assay reagent and research-grade chemical. It is commonly applied in metabolic studies, glycosylation research, and enzymatic assays, supplied as a high-purity crystalline powder suitable for laboratory use.
  • High purity suitable for analytical and biochemical assays.
  • Well-characterized chemical properties (C8H15NO6; 221.21 g/mol).
  • Relevant for studies of glycosylation, chondrocyte biology, and metabolism.
  • Available in lab-scale quantities for bench-top experiments.
  • Documentation available: certificate of analysis and safety data sheet.
Catalog No. 50-002-82239 Supplier Medchemexpress LLC Supplier No. HYW2715171G
N-Acetyl-α-D-glucosamine is an amino sugar used as a biochemical assay reagent and research-grade chemical. It is commonly applied in metabolic studies, glycosylation research, and enzymatic assays, supplied as a high-purity crystalline powder suitable for laboratory use.
  • High purity suitable for analytical and biochemical assays.
  • Well-characterized chemical properties (C8H15NO6; 221.21 g/mol).
  • Relevant for studies of glycosylation, chondrocyte biology, and metabolism.
  • Available in lab-scale quantities for bench-top experiments.
  • Documentation available: certificate of analysis and safety data sheet.