Learn More
Alkali Scientific N Acetyl D glucosamine [2 Acetamido 2 deoxy D glucopyranose GlcNAc], 1 Kilogram
SDP
Catalog No. 5000661270
Encompass_Preferred
N-Acetyl D-Glucosamine is an amino sugar derived from glucosamine, used in a variety of biochemical research applications, particularly in carbohydrate metabolism studies. This 1 kg bottle of N-Acetyl D-Glucosamine is utilized for exploring cell wall biosynthesis, glycolysis, and glycosylation in both eukaryotic and prokaryotic organisms. It plays a key role in the production of chitin and is widely used in studies of glycosaminoglycans and proteoglycans. This reagent is critical for biochemical assays in cell biology, molecular genetics, and structural biology, where it serves as a substrate for enzymatic reactions or as an analytical tool in metabolic studies.
Catalog No. 50-006-61270 Supplier Alkali Scientific Supplier No. NAC002CP
N-Acetyl D-Glucosamine is an amino sugar derived from glucosamine, used in a variety of biochemical research applications, particularly in carbohydrate metabolism studies. This 1 kg bottle of N-Acetyl D-Glucosamine is utilized for exploring cell wall biosynthesis, glycolysis, and glycosylation in both eukaryotic and prokaryotic organisms. It plays a key role in the production of chitin and is widely used in studies of glycosaminoglycans and proteoglycans. This reagent is critical for biochemical assays in cell biology, molecular genetics, and structural biology, where it serves as a substrate for enzymatic reactions or as an analytical tool in metabolic studies.