Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Acetyl-D-Glucosamine, MP Biomedicals™

Catalog No. MP210006810 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP210006810 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP210006810 Supplier MP Biomedicals, Inc Supplier No. 0210006810
Add to Cart
Add to Cart

Used to identify, differentiate and characterize N-acetyl-β-D-hexoaminidase(s). N-Acetyl-D-Glucosamine, MP Biomedicals is a derivitized glucose monomer found in polymers of bacterial cell walls, chitin, hyaluronic acids and various glycans.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7512-17-6
Molecular Formula C8H15NO6
Molecular Weight (g/mol) 221.21
MDL Number MFCD00061615
InChI Key OVRNDRQMDRJTHS-WZWLWIOANA-N
Synonym 2-Acetamido-2-deoxy-D-glucose, D-GlcNAc
IUPAC Name N-[(3R,4R,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide
SMILES CC(=O)N[C@H]1C(O)O[C@H](CO)[C@@H](O)[C@@H]1O

Specifications

Melting Point 199°C (literature)
pH 7.20 (literature)
Quantity 10 g
Solubility Information Soluble in water (50mg/mL).
Formula Weight 221.21
Physical Form Powder
Recommended Storage +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.