Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diazyme Laboratories N-Acetyl-β-D-Glucosaminidase Calibrator

Catalog No. 23700575
Encompass
Encompass_Preferred
Change view
Click to view available options
Format:
1 x 2 mL Vial
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
23-700-575 1 x 2 mL Vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-700-575 Supplier Diazyme Laboratories Supplier No. DZ062ACAL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

N-Acetyl-β-D-glucosaminidase (NAG) Calibrator Set is for determination of NAG in urine samples.

  • Ready-to-use for manual methods and adaptable for analyzers
  • The assay is specific for NAG and has  excellent sensitivity with a linear range up to 200 IU/L2 and excellent precision with a CV% of <5%
  • There is no significant interference from hemoglobin or albumin
  • Interference from bilirubin occurs only at levels higher than 5 mg/dL

Specifications

Certifications/Compliance USA: For Research Use Only; EU: CE IVD
Content And Storage Stored at -20°C; Store CALIBRATOR at 2°C to 8°C after reconstitution; DO NOT FREEZE LIQUID MATERIAL; Note: Store CALIBRATOR tightly capped when not in use
Description N-Acetyl-β-D-Glucosaminidase (NAG) Assay Calibrator Set
Concentration Lab dependent
Format 1 x 2 mL Vial
Type Diagnostic
Includes Lyophilized CALIBRATOR 1 (REF DZ062A-S1V)
Stability Unconstituted material: stability is up to the expiration date on label when stored at -20°C; Constituted material: stability is up to 14 days when stored at 2 to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.