Learn More
Medchemexpress LLC HY-128850 100mg Medchemexpress, N-Acetyl-D-mannosamine CAS:3615-17-6 Purity:>98%
SDP
Catalog No. 501932638
Encompass_Preferred
Medchemexpress, HY-128850 100mg N-Acetyl-D-mannosamine CAS:3615-17-6 N-Acetyl-D-mannosamine (ManNAc) is an essential precursor of N-acetylneuraminic acid (NeuAc), the specific monomer of bacterial capsular polysialic acid (PA) [1] .N-Acetyl-D-mannosamine (ManNAc) can be metabolized by GNE and GlcNAc 2-epimerase (Renin binding protein, RnBP), into ManNAc-6-phosphate and GlcNAc, respectively. N-Acetyl-d-mannosamine (ManNAc) and its derivatives activates hypocretin (HCRT) gene expression in the orexin neurons, providing a potential model for the testing of a therapy for neural disorders [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2638 Supplier Medchemexpress LLC Supplier No. HY128850100MG
Medchemexpress, HY-128850 100mg N-Acetyl-D-mannosamine CAS:3615-17-6 N-Acetyl-D-mannosamine (ManNAc) is an essential precursor of N-acetylneuraminic acid (NeuAc), the specific monomer of bacterial capsular polysialic acid (PA) [1] .N-Acetyl-D-mannosamine (ManNAc) can be metabolized by GNE and GlcNAc 2-epimerase (Renin binding protein, RnBP), into ManNAc-6-phosphate and GlcNAc, respectively. N-Acetyl-d-mannosamine (ManNAc) and its derivatives activates hypocretin (HCRT) gene expression in the orexin neurons, providing a potential model for the testing of a therapy for neural disorders [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.