Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Acetyl-β-D-mannosamine Hydrate, MP Biomedicals™

Catalog No. 021001092 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02100109-2 250 mg
ICN10010901 1 g
ICN10010905 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 02100109-2 Supplier MP Biomedicals, Inc Supplier No. 02100109.2
Add to Cart
Add to Cart

N-Acetyl-β-D-mannosamine Hydrate is an essential precursor of N-acetylneuraminic acid (NeuAc).

  • N-Acetyl-β-D-mannosamine hydrate is used as a substrate to identify, differentiate and characterize enzymes such as N-acetyl-glucosamine epimerase(s) and aldolase(s).
  • N-Acetyl-D-mannosamine is used for the synthesis of of N-acetylneuraminic acid.
TRUSTED_SUSTAINABILITY

Specifications

Color White to Off-white
Form Crystalline
Quantity 250 mg
Product Type N-acetyl-β-D-Mannosamine
Purity or Quality Grade >98%

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.