Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Acetyl-D-mannosamine monohydrate, 99%

Catalog No. AAL11167MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL11167MD 250 mg
AAL1116703 1 g
AAL1116706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL11167MD Supplier Thermo Scientific Chemicals Supplier No. L11167MD

missing translation for 'validMainframeMsg'

CAS: 1071625-31-4 | C8H17NO7 | 239.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1071625-31-4
Molecular Formula C8H17NO7
Molecular Weight (g/mol) 239.22
MDL Number MFCD00149493,MFCD00046758,MFCD00149493,MFCD00069808
InChI Key VVQPUTSNIMAJPT-UHFFFAOYNA-N
Synonym n-acetylmannosamine
PubChem CID 11908605
IUPAC Name N-[(2R,3S,4S,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide
SMILES O.CC(=O)NC1C(O)OC(CO)C(O)C1O

Specifications

Melting Point ∼130°C (decomposition)
Packaging Glass bottle
Quantity 250 mg
Beilstein 1346524
Formula Weight 239.24 (221.22 anhydrous)
Percent Purity 99%
Physical Form Powder
Chemical Name or Material N-Acetyl-D-mannosamine monohydrate
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.