Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-acetyl-D-valine | 17916-88-0 | MFCD00026457; MFCD03939895 | 97.0% | 159.18 g/mol | C7H13NO3 | 100 G
SDP

Catalog No. 5000302551
Encompass_Preferred

N-acetyl-D-valine is the N-acetylated derivative of the amino acid D-valine provided for research use. It is intended as a synthetic building block and analytical reagent for medicinal chemistry, peptide synthesis, and biochemical studies.

  • High chemical purity (97.0%).
  • Molecular formula C7H13NO3; molecular weight 159.18 g/mol.
  • Solid appearance; hygroscopic considerations for solvents.
  • Soluble in DMSO at approximately 100 mg/mL; may require sonication.
  • Recommended storage: powder at -20°C (long term) or 4°C (short term); in solvent at -80°C or -20°C as specified.
  • Suitable as a synthetic building block and reference standard for research applications.

Catalog No. 50-003-02551 Supplier Medchemexpress LLC Supplier No. HYW015465100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-acetyl-D-valine is the N-acetylated derivative of the amino acid D-valine provided for research use. It is intended as a synthetic building block and analytical reagent for medicinal chemistry, peptide synthesis, and biochemical studies.

  • High chemical purity (97.0%).
  • Molecular formula C7H13NO3; molecular weight 159.18 g/mol.
  • Solid appearance; hygroscopic considerations for solvents.
  • Soluble in DMSO at approximately 100 mg/mL; may require sonication.
  • Recommended storage: powder at -20°C (long term) or 4°C (short term); in solvent at -80°C or -20°C as specified.
  • Suitable as a synthetic building block and reference standard for research applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.