Learn More
Medchemexpress LLC N-(Acetyl-d3)-S-benzyl-L-cysteine | 201404-15-1 | 256.34 | 1 MG
SDP
Catalog No. 5000349488
Encompass_Preferred
N-(Acetyl-d3)-S-benzyl-L-cysteine is the deuterium labeled N-(Acetyl)-S-benzyl-L-cysteine, intended for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-49488 Supplier Medchemexpress LLC Supplier No. HY139491S1MG
N-(Acetyl-d3)-S-benzyl-L-cysteine is the deuterium labeled N-(Acetyl)-S-benzyl-L-cysteine, intended for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.