Learn More
Medchemexpress LLC N-Acetyl dapsone-d4 | 2070015-08-4 | 98.0% | 294.36 | 5 MG
SDP
Catalog No. 5000431820
Encompass_Preferred
N-acetyl dapsone-d4 is a deuterium-labeled N-acetyl Dapsone, a metabolite of Dapsone. It is intended for research use only and not for patient use. This compound is valuable as a tracer and as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-31820 Supplier Medchemexpress LLC Supplier No. HYG0016S5MG
N-acetyl dapsone-d4 is a deuterium-labeled N-acetyl Dapsone, a metabolite of Dapsone. It is intended for research use only and not for patient use. This compound is valuable as a tracer and as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS