Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-acetyl dapsone-d4 | 2070015-08-4 | MFCD28900735 | 98.0% | 294.36 | C14H10D4N2O3S | 10 MG
SDP

Catalog No. 5000297503
Encompass_Preferred

N-Acetyl dapsone-d4 is the deuterium-labeled form of N-acetyl dapsone, the primary metabolite of dapsone. It is supplied as an isotope-labeled analytical standard for research and quantitative analytical applications.

  • Deuterium-labeled internal standard for LC-MS and GC-MS.
  • Suitable for quantitative and metabolic studies.
  • Molecular formula C14H10D4N2O3S; molecular weight 294.36.
  • Packaged as a 10 mg vial for laboratory use.
  • Intended for research use only; not for human or veterinary use.

Catalog No. 50-002-97503 Supplier Medchemexpress LLC Supplier No. HYG0016S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Acetyl dapsone-d4 is the deuterium-labeled form of N-acetyl dapsone, the primary metabolite of dapsone. It is supplied as an isotope-labeled analytical standard for research and quantitative analytical applications.

  • Deuterium-labeled internal standard for LC-MS and GC-MS.
  • Suitable for quantitative and metabolic studies.
  • Molecular formula C14H10D4N2O3S; molecular weight 294.36.
  • Packaged as a 10 mg vial for laboratory use.
  • Intended for research use only; not for human or veterinary use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.