Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Acetyl-DL-histidine Monohydrate 98.0+%, TCI America™
SDP

Catalog No. A06981G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A06981G 1 g
A06985G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A06981G Supplier TCI America Supplier No. A06981G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 213178-97-3
Molecular Formula C8H13N3O4
Molecular Weight (g/mol) 215.209
MDL Number MFCD00065961
InChI Key PSWSDQRXCOJSFC-UHFFFAOYSA-N
Synonym 2-acetamido-3-1h-imidazol-4-yl propanoic acid hydrate, ac-dl-his-oh.h2o, n-acetyl-dl-histidine monohydrate, 2-acetamido-3-1h-imidazol-5-yl propanoic acid hydrate, 2-acetylamino-3-1h-imidazol-4-yl propanoic acid hydrate, n-acetyl-l-histidine monohydrate, 2-acetamido-3-3h-imidazol-4-yl propanoic acid hydrate, acmc-1c6iz, acmc-209j3m, n-acetyl-dl-histidine hydrate
PubChem CID 18668507
IUPAC Name 2-acetamido-3-(1H-imidazol-5-yl)propanoic acid;hydrate
SMILES CC(=O)NC(CC1=CN=CN1)C(=O)O.O

Specifications

Melting Point 152°C
Color White
Quantity 1 g
Formula Weight 197.19
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-Acetyl-DL-histidine Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.