Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-acetyl-DL-methionine | 1115-47-5 | MFCD00008681 | 98.7% | 191.25 g/mol | C7H13NO3S | 500 MG
SDP

Catalog No. 5000300973
Encompass_Preferred

N-acetyl-DL-methionine is the N-acetylated derivative of methionine used as a biochemical reagent and endogenous metabolite reference. Supplied as a white to off-white powder, it is intended for research and analytical applications, including metabolic studies and method development. Not for human therapeutic use.

  • Molecular formula: C7H13NO3S.
  • Molecular weight: 191.25 g/mol.
  • Purity: 98.7%.
  • Appearance: white to off-white powder.
  • Applications include metabolic research and analytical reference standards.
  • Solubility: soluble in water and DMSO.

Catalog No. 50-003-00973 Supplier Medchemexpress LLC Supplier No. HYW019704500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-acetyl-DL-methionine is the N-acetylated derivative of methionine used as a biochemical reagent and endogenous metabolite reference. Supplied as a white to off-white powder, it is intended for research and analytical applications, including metabolic studies and method development. Not for human therapeutic use.

  • Molecular formula: C7H13NO3S.
  • Molecular weight: 191.25 g/mol.
  • Purity: 98.7%.
  • Appearance: white to off-white powder.
  • Applications include metabolic research and analytical reference standards.
  • Solubility: soluble in water and DMSO.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.