Learn More
Enzo Life Sciences N-Acetyl-eglin C, (recombinant) (0.5mg). CAS: 96380-69-7
SDP
Supplier:  Enzo Life Sciences ALX201006MC05
Encompass_Preferred
Protease inhibitor. Eglin C was originally isolated from the leech Hirudo medicinalis . Recombinant N-acetyl-eglin C is a 70 amino acid peptide with identical biological activity to native eglin C. An effective inhibitor of chymotrypsin and subtilisin as well as of leukocyte elastase and cathepsin G. Formula: C377H552N96O107. MW: 8141.1. Purity: ≥97% (HPLC). Appearance: White to off-white powder. Solubility: Soluble in water. Long Term Storage: -20°C.
Catalog No. 50-200-7887