Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-acetyl-glucosamine thiazoline | 179030-22-9 | MFCD22376563 | >95.0% | C8H13NO4S | 10MG
SDP

Catalog No. 5000260427
Encompass_Preferred

N-acetyl-glucosamine thiazoline (NAG-thiazoline) is a small-molecule inhibitor of O-GlcNAcase used in biochemical and enzymology research to probe O-GlcNAc removal and GH20 family GlcNAcase activity. It shows potent biochemical inhibition against O-GlcNAcase and measurable activity versus bacterial GH20 GlcNAcase.

  • Selective O-GlcNAcase inhibition for mechanistic studies.
  • Documented biochemical potency (Ki ≈ 180 nM) against O-GlcNAcase.
  • Activity data available versus GH20 GlcNAcase (IC50 ≈ 11.9 μM).
  • Suitable for enzymology assays and pathway probing.
  • Available as solid and in solution formats for assay flexibility.
  • Molecular formula C8H13NO4S; CAS 179030-22-9.

Catalog No. 50-002-60427 Supplier Medchemexpress LLC Supplier No. HY13943110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

N-acetyl-glucosamine thiazoline (NAG-thiazoline) is a small-molecule inhibitor of O-GlcNAcase used in biochemical and enzymology research to probe O-GlcNAc removal and GH20 family GlcNAcase activity. It shows potent biochemical inhibition against O-GlcNAcase and measurable activity versus bacterial GH20 GlcNAcase.

  • Selective O-GlcNAcase inhibition for mechanistic studies.
  • Documented biochemical potency (Ki ≈ 180 nM) against O-GlcNAcase.
  • Activity data available versus GH20 GlcNAcase (IC50 ≈ 11.9 μM).
  • Suitable for enzymology assays and pathway probing.
  • Available as solid and in solution formats for assay flexibility.
  • Molecular formula C8H13NO4S; CAS 179030-22-9.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.