Learn More
Cayman Chemical N-Acetyl-L-alanine, Purity: ≥95%, CAS: 97-69-8, 25 g
SDP
Catalog No. 502917777
Encompass_Preferred
An endogenous metabolite; decreases IFN-γ production induced by radiation-killed M. tuberculosis in NK cells isolated from patients with latent M. tuberculosis infection; decreases ATP production and glycolytic flux in naïve or radiation-killed M. tuberculosis-exposed isolated human NK cells; brain levels are decreased in a rat model of STZ-induced diabetic neuropathic pain; increased urine levels are associated with mutations in ACY1
Catalog No. 50-291-7777 Supplier Cayman Chemical Supplier No. 3736025 G
An endogenous metabolite; decreases IFN-γ production induced by radiation-killed M. tuberculosis in NK cells isolated from patients with latent M. tuberculosis infection; decreases ATP production and glycolytic flux in naïve or radiation-killed M. tuberculosis-exposed isolated human NK cells; brain levels are decreased in a rat model of STZ-induced diabetic neuropathic pain; increased urine levels are associated with mutations in ACY1