Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-acetyl-L-asparagine | 4033-40-3 | MFCD00066023 | 98.0% | 174.15 g/mol | C6H10N2O4 | 1 ML
SDP

Catalog No. 5000301014
Encompass_Preferred

(S)-2-acetamido-4-amino-4-oxobutanoic acid, also known as N-acetyl-L-asparagine, is an endogenous metabolite supplied for laboratory research. It is available as a solid and as a ready-to-use 10 mM solution in DMSO, with high purity and defined storage recommendations to support reproducible metabolic and biochemical studies. For research use only.

  • Available as solid and 10 mM solution in DMSO.
  • High purity suitable for analytical and biochemical assays.
  • Molecular formula C6H10N2O4; molecular weight 174.15 g/mol.
  • Storage guidelines provided for powder and solution to preserve stability.
  • Intended for research use only; not for clinical or human use.

Catalog No. 50-003-01014 Supplier Medchemexpress LLC Supplier No. HYW01679810MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(S)-2-acetamido-4-amino-4-oxobutanoic acid, also known as N-acetyl-L-asparagine, is an endogenous metabolite supplied for laboratory research. It is available as a solid and as a ready-to-use 10 mM solution in DMSO, with high purity and defined storage recommendations to support reproducible metabolic and biochemical studies. For research use only.

  • Available as solid and 10 mM solution in DMSO.
  • High purity suitable for analytical and biochemical assays.
  • Molecular formula C6H10N2O4; molecular weight 174.15 g/mol.
  • Storage guidelines provided for powder and solution to preserve stability.
  • Intended for research use only; not for clinical or human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.